Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gex-3
Entrez Gene ID
178191
Gene Name
Membrane-associated protein gex-3
Gene Aliases
CELE_F28D1.10
Chromosome Location
Chr.IV: 12404431 - 12408697 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069967.6 NP_502368.1 9 - 10 2274 126
GenBank mRNA AB073210.1 - 9 - 10 2270 126

View Full Product Details