Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
him-6
Entrez Gene ID
178201
Gene Name
Bloom syndrome protein homolog
Gene Aliases
CELE_T04A11.6
Chromosome Location
Chr.IV: 12490791 - 12495298 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069989.7 NP_502390.2 3 - 4 1054 109
GenBank mRNA AY095296.1 - 3 - 4 1076 109

View Full Product Details