Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-18
Entrez Gene ID
188047
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_T04A11.9
Chromosome Location
Chr.IV: 12502256 - 12504126 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069992.3 NP_502393.2 1 - 2 292 114

View Full Product Details