Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-13
Entrez Gene ID
186151
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_F53B2.4
Chromosome Location
Chr.IV: 12521239 - 12522621 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069996.2 NP_502397.2 2 - 3 440 107

View Full Product Details