Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-18
Entrez Gene ID
178237
Gene Name
Receptor-type guanylate cyclase gcy-18
Gene Aliases
CELE_ZK896.8
Chromosome Location
Chr.IV: 12860097 - 12865956 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070048.7 NP_502449.2 13 - 14 2413 69
GenBank mRNA AB201389.1 - 13 - 14 2373 69

View Full Product Details