Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scl-9
Entrez Gene ID
186048
Gene Name
SCP domain-containing protein
Gene Aliases
CELE_F49E11.4
Chromosome Location
Chr.IV: 13047148 - 13048266 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070096.3 NP_502497.1 2 - 3 230 86

View Full Product Details