Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scl-11
Entrez Gene ID
186050
Gene Name
SCP domain-containing protein
Gene Aliases
CELE_F49E11.6
Chromosome Location
Chr.IV: 13051373 - 13052219 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070098.2 NP_502499.1 2 - 3 320 100

View Full Product Details