Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cho-1
Entrez Gene ID
178274
Gene Name
High-affinity choline transporter 1
Gene Aliases
CELE_C48D1.3
Chromosome Location
Chr.IV: 13209729 - 13214566 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070138.8 NP_502539.1 6 - 7 1022 120
GenBank mRNA AB030946.1 - 6 - 7 1023 120

View Full Product Details