Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-25
Entrez Gene ID
190453
Gene Name
protein-coding
Gene Aliases
CELE_Y62E10A.4
Chromosome Location
Chr.IV: 13365169 - 13368435 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268803.2 NP_001255732.1 5 - 6 709 64
NM_001268804.3 NP_001255733.1 1 - 2 100 64

View Full Product Details