Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-48
Entrez Gene ID
188814
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_T23G4.4
Chromosome Location
Chr.IV: 13673804 - 13677886 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070242.2 NP_502643.1 7 - 8 1265 104

View Full Product Details