Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
oac-56
Entrez Gene ID
190505
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_Y67A10A.1
Chromosome Location
Chr.IV: 14345383 - 14350751 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070341.3 NP_502742.1 3 - 4 395 105

View Full Product Details