Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ssu-2
Entrez Gene ID
178379
Gene Name
protein-coding
Gene Aliases
CELE_Y67A10A.6
Chromosome Location
Chr.IV: 14361247 - 14368322 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268875.3 NP_001255804.1 5 - 6 873 77
NM_001268876.2 NP_001255805.1 5 - 6 863 77
NM_001268877.3 NP_001255806.1 5 - 6 956 77
NM_001306937.4 NP_001293866.1 5 - 6 1059 77

View Full Product Details