Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srt-44
Entrez Gene ID
187500
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_M199.1
Chromosome Location
Chr.IV: 15103629 - 15106483 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070422.1 NP_502823.1 3 - 4 712 77

View Full Product Details