Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pkd-2
Entrez Gene ID
178424
Gene Name
Polycystin-2
Gene Aliases
CELE_Y73F8A.1
Chromosome Location
Chr.IV: 15227814 - 15234993 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070437.5 NP_502838.3 4 - 5 889 74

View Full Product Details