Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-25
Entrez Gene ID
191653
Gene Name
protein-coding
Gene Aliases
CELE_Y105C5B.2
Chromosome Location
Chr.IV: 15872933 - 15882377 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307187.2 NP_001294116.1 18 - 19 2990 107
NM_001307188.3 NP_001294117.1 17 - 18 2804 107

View Full Product Details