Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
jac-1
Entrez Gene ID
178451
Gene Name
protein-coding
Gene Aliases
CELE_Y105C5B.21
Chromosome Location
Chr.IV: 15997835 - 16026847 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047542.3 NP_001041007.1 11 - 12 3158 78
NM_001047543.4 NP_001041008.1 8 - 9 2648 78
NM_001268952.3 NP_001255881.1 9 - 10 2939 78
NM_001268953.4 NP_001255882.1 7 - 8 2453 78

View Full Product Details