Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-61
Entrez Gene ID
186783
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_H25K10.7
Chromosome Location
Chr.IV: 16236732 - 16238655 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268956.1 NP_001255885.1 3 - 4 969 162
NR_066102.1 3 - 4 901 162

View Full Product Details