Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snr-1
Entrez Gene ID
178483
Gene Name
Small nuclear ribonucleoprotein Sm D3
Gene Aliases
CELE_Y116A8C.42
Chromosome Location
Chr.IV: 17088058 - 17088946 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070626.6 NP_503027.1 1 - 2 78 63

View Full Product Details