Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
elo-2
Entrez Gene ID
178532
Gene Name
Elongation of very long chain fatty acids protein
Gene Aliases
CELE_F11E6.5
Chromosome Location
Chr.IV: 17466062 - 17468458 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070713.8 NP_503114.1 5 - 6 949 144

View Full Product Details