Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ife-3
Entrez Gene ID
178536
Gene Name
protein-coding
Gene Aliases
CELE_B0348.6
Chromosome Location
Chr.V: 6584 - 7829 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070722.5 NP_503123.1 2 - 3 348 64

View Full Product Details