Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp-29A3
Entrez Gene ID
189649
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_Y38C9B.1
Chromosome Location
Chr.V: 59239 - 63633 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070729.7 NP_503130.2 6 - 7 1200 61

View Full Product Details