Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpn-1
Entrez Gene ID
177455
Gene Name
26S proteasome non-ATPase regulatory subunit 2
Gene Aliases
CELE_T22D1.9
Chromosome Location
Chr.IV: 6906370 - 6909913 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068663.5 NP_501064.1 4 - 5 2616 68

View Full Product Details