Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
him-3
Entrez Gene ID
177462
Gene Name
HORMA domain-containing protein
Gene Aliases
CELE_ZK381.1
Chromosome Location
Chr.IV: 6977519 - 6978943 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068677.6 NP_501078.1 1 - 2 207 72
NR_131562.1 1 - 2 200 72
GenBank mRNA AF172351.1 - 1 - 2 218 72

View Full Product Details