Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kin-34
Entrez Gene ID
178552
Gene Name
Protein kinase domain-containing protein
Gene Aliases
CELE_R02C2.2
Chromosome Location
Chr.V: 244407 - 246197 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070761.7 NP_503162.3 3 - 4 752 89

View Full Product Details