Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-62
Entrez Gene ID
191867
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_ZK6.9
Chromosome Location
Chr.V: 401688 - 403128 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070789.1 NP_503190.1 2 - 3 557 87

View Full Product Details