Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-98
Entrez Gene ID
178568
Gene Name
protein-coding
Gene Aliases
CELE_M02H5.6
Chromosome Location
Chr.V: 452207 - 456958 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313226.3 NP_001300155.1 4 - 5 456 76
NM_001372992.2 NP_001360400.1 9 - 9 1303 76
NR_132478.1 4 - 5 381 76

View Full Product Details