Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-99
Entrez Gene ID
178569
Gene Name
Nuclear Hormone Receptor family
Gene Aliases
CELE_M02H5.1
Chromosome Location
Chr.V: 474778 - 477155 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070813.3 NP_503214.1 7 - 8 1191 85

View Full Product Details