Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- srj-53
- Entrez Gene ID
-
191952
- Gene Name
- Serpentine Receptor, class J
- Gene Aliases
-
CELE_Y49C4A.6
- Chromosome Location
-
Chr.V: 693541 - 694725 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070896.3 |
NP_503297.3 |
2 - 3 |
555 |
132 |
- Gene Symbol
- srj-52
- Entrez Gene ID
-
13211474
- Gene Name
- Serpentine Receptor, class J
- Gene Aliases
-
CELE_Y49C4A.7
- Chromosome Location
-
Chr.V: 695326 - 696014 on Build WBcel235
View Full Product Details