Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-38
Entrez Gene ID
3565932
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_Y49C4A.2
Chromosome Location
Chr.V: 707218 - 708453 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070900.2 NP_503301.2 3 - 4 515 150

View Full Product Details