Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-24
Entrez Gene ID
191942
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_C05E4.11
Chromosome Location
Chr.V: 736803 - 738870 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070908.4 NP_503309.1 3 - 4 352 71

View Full Product Details