Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-20
Entrez Gene ID
191970
Gene Name
protein-coding
Gene Aliases
CELE_C05E4.2
Chromosome Location
Chr.V: 756469 - 757892 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313121.3 NP_001300050.1 2 - 3 521 134
NM_070915.4 NP_503316.1 2 - 3 521 134

View Full Product Details