Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- sqv-6
- Entrez Gene ID
-
190099
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_Y50D4C.4
- Chromosome Location
-
Chr.V: 950306 - 964789 on Build WBcel235
- Gene Symbol
- Y50D4C.6
- Entrez Gene ID
-
178600
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_Y50D4C.6
- Chromosome Location
-
Chr.V: 946723 - 964797 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_171435.5 |
NP_741508.2 |
4 - 5 |
1507 |
88 |
View Full Product Details