Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ergo-1
Entrez Gene ID
178602
Gene Name
protein-coding
Gene Aliases
CELE_R09A1.1
Chromosome Location
Chr.V: 1007317 - 1018358 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313235.3 NP_001300164.1 4 - 5 2039 67
NM_001313236.3 NP_001300165.1 3 - 4 1196 67
NM_001313237.3 NP_001300166.1 1 - 2 827 67
NM_070961.9 NP_503362.2 7 - 8 3456 67

View Full Product Details