Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-67
Entrez Gene ID
191984
Gene Name
Seven TM Receptor
Gene Aliases
CELE_K10C9.6
Chromosome Location
Chr.V: 1062588 - 1064591 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070970.1 NP_503371.1 2 - 3 941 140

View Full Product Details