Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-31
Entrez Gene ID
178643
Gene Name
NRF domain-containing protein
Gene Aliases
CELE_F52F10.3
Chromosome Location
Chr.V: 1538354 - 1541886 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071040.5 NP_503441.2 1 - 2 363 101

View Full Product Details