Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-23
Entrez Gene ID
183490
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_C45H4.9
Chromosome Location
Chr.V: 2153574 - 2155799 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071182.3 NP_503583.2 5 - 6 733 79

View Full Product Details