Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- cyp-33C1
- Entrez Gene ID
-
183484
- Gene Name
- CYtochrome P450 family
- Gene Aliases
-
CELE_C45H4.2
- Chromosome Location
-
Chr.V: 2179514 - 2181938 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071191.3 |
NP_503592.2 |
5 - 6 |
1466 |
91 |
- Gene Symbol
- cyp-33C2
- Entrez Gene ID
-
178695
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C45H4.17
- Chromosome Location
-
Chr.V: 2181366 - 2186599 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071193.3 |
NP_503594.1 |
1 - 2 |
249 |
91 |
View Full Product Details