Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-33C7
Entrez Gene ID
185589
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_F41B5.2
Chromosome Location
Chr.V: 2250540 - 2252591 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071217.3 NP_503618.2 2 - 3 709 132

View Full Product Details