Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- nas-2
- Entrez Gene ID
-
186345
- Gene Name
- Zinc metalloproteinase nas-2
- Gene Aliases
-
CELE_F56A4.1
- Chromosome Location
-
Chr.V: 2466762 - 2468504 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071277.3 |
NP_503678.3 |
3 - 3 |
299 |
148 |
- Gene Symbol
- Y19D10A.6
- Entrez Gene ID
-
189484
- Gene Name
- Metalloendopeptidase
- Gene Aliases
-
CELE_Y19D10A.6
- Chromosome Location
-
Chr.V: 2358801 - 2360543 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071251.3 |
NP_503652.3 |
3 - 3 |
299 |
148 |
View Full Product Details