Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-20
Entrez Gene ID
191940
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_Y45G12C.15
Chromosome Location
Chr.V: 2571527 - 2573208 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071303.1 NP_503704.1 3 - 4 353 141

View Full Product Details