Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acr-17
Entrez Gene ID
191603
Gene Name
protein-coding
Gene Aliases
CELE_F53E10.2
Chromosome Location
Chr.V: 2608652 - 2615262 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028790.6 NP_001023961.1 2 - 3 141 106
NM_001316802.4 NP_001303731.1 2 - 3 145 106
NR_133607.1 2 - 3 141 106
NR_133608.1 2 - 3 141 106

View Full Product Details