Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-29
Entrez Gene ID
182839
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_C24B9.5
Chromosome Location
Chr.V: 2717395 - 2719135 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071336.2 NP_503737.2 2 - 3 151 142

View Full Product Details