Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ketn-1
Entrez Gene ID
178740
Gene Name
protein-coding
Gene Aliases
CELE_F54E2.3
Chromosome Location
Chr.V: 2781939 - 2807860 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307737.5 NP_001294666.1 14 - 15 4667 66
NM_001307738.4 NP_001294667.1 14 - 15 4667 66
NM_001307739.4 NP_001294668.1 14 - 15 4667 66
NM_001307740.4 NP_001294669.1 14 - 15 4667 66
NM_001316807.3 NP_001303736.1 1 - 2 191 66
NM_001377590.2 NP_001364781.1 6 - 7 2750 66
NM_001377591.2 NP_001364828.1 6 - 7 2750 66
NM_001377592.1 NP_001364782.1 6 - 7 2750 66
NM_001377593.1 NP_001364783.1 6 - 7 2750 66
NM_001377594.2 NP_001364827.1 6 - 7 2750 66
GenBank mRNA AY819766.1 - 9 - 10 3537 66

View Full Product Details