Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ketn-1
Entrez Gene ID
178740
Gene Name
protein-coding
Gene Aliases
CELE_F54E2.3
Chromosome Location
Chr.V: 2781939 - 2807860 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307737.5 NP_001294666.1 17 - 18 5182 63
NM_001307738.4 NP_001294667.1 17 - 18 5182 63
NM_001307739.4 NP_001294668.1 17 - 18 5182 63
NM_001307740.4 NP_001294669.1 17 - 18 5182 63
NM_001316807.3 NP_001303736.1 4 - 5 706 63
NM_001316808.3 NP_001303737.1 1 - 2 295 63
NM_001377590.2 NP_001364781.1 9 - 10 3265 63
NM_001377591.2 NP_001364828.1 9 - 10 3265 63
NM_001377592.1 NP_001364782.1 9 - 10 3265 63
NM_001377593.1 NP_001364783.1 9 - 10 3265 63
NM_001377594.2 NP_001364827.1 9 - 10 3265 63
GenBank mRNA AY819766.1 - 12 - 13 4052 63

View Full Product Details