Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-46
Entrez Gene ID
191979
Gene Name
Seven TM Receptor
Gene Aliases
CELE_C31B8.6
Chromosome Location
Chr.V: 2898297 - 2899967 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071387.2 NP_503788.1 3 - 4 926 80
NR_133614.1 4 - 5 721 80

View Full Product Details