Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tat-6
Entrez Gene ID
178754
Gene Name
Phospholipid-transporting ATPase
Gene Aliases
CELE_F02C9.3
Chromosome Location
Chr.V: 3129103 - 3133412 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071457.2 NP_503858.1 6 - 7 2622 89

View Full Product Details