Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-16
Entrez Gene ID
191969
Gene Name
Seven TM Receptor
Gene Aliases
CELE_Y73C8A.1
Chromosome Location
Chr.V: 3236875 - 3240611 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377595.2 NP_001364843.1 3 - 4 760 107
NR_134547.1 5 - 6 1127 107

View Full Product Details