Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-11
Entrez Gene ID
189018
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_T28A11.12
Chromosome Location
Chr.V: 3241424 - 3242636 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071487.1 NP_503888.1 4 - 5 751 102

View Full Product Details