Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srbc-30
Entrez Gene ID
182096
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_C02A12.3
Chromosome Location
Chr.V: 3478031 - 3480446 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071569.2 NP_503970.2 2 - 3 555 156

View Full Product Details