Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-67
Entrez Gene ID
188827
Gene Name
protein-coding
Gene Aliases
CELE_T24A6.10
Chromosome Location
Chr.V: 3538897 - 3541217 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322533.3 NP_001309659.1 1 - 2 169 143
NM_071588.2 NP_503989.1 1 - 2 1057 143

View Full Product Details